Learn More
Bel-Art™ Mercury Collector
Product Code. 10595444
Change view
Click to view available options
For Use With (Application):
Mercury spill clean-up
Mercury spill clean-up replacement pad
Unit Size:
Each
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10595444 Mercury spill clean-up Each
12372088 Mercury spill clean-up replacement pad Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10595444 Supplier Bel-Art™ Supplier No. F362500000
  • ONLINE EXCLUSIVE PRICE
    305.00 DKK
    198.88 DKK / Pakke med 1
    Spare 106.12 DKK
    34% Slukket
Forventet forsendelse: 23-12-2026
Captures spilled mercury beads quickly and conveniently. Bel-Art™ Mercury Collector holds 125 ml of mercury.
  • Lid features absorbent foam pad on interior surface
  • Replacement foam pad available
  • Dispose of mercury by approved disposal method
Specifications
Color White
Capacity (English) 4.22 oz.
Capacity (Metric) 125 mL
Material Polyethylene
Type Mercury Collector
For Use With (Application) Mercury spill clean-up
Includes Jar, perforated plate, lid with attached foam pad
Height (English) 1.88 in.
Height (Metric) 4.77 cm