missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Double Spinfin™ Magnetic Stirring Bars

To agitate liquids in round bottom vessels where other stirring bars cannot be used.

Brand:  Bel-Art™ F37126-0034

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15755872

  • 166.00 DKK / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket

Description

Description

  • Made from PTFE
  • Fins on top and bottom ensure complete agitation
  • Does not wobble, maintains position in vessel
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclavable
Specifications

Specifications

19 mm
16mm
50
Alnico Magnet
Cylinder
0.75 in.
White
0.62 in.
FDA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers