Få mere at vide
Beclamide, TRC
Artikelnummer. 30341703
Skift visning
Mængde:
2.5 g
Artikelnummer. Mængde
30341703 2.5 g
1 options
Artikelnummer. 30341703 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1194002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Beclamide
CAS 501-68-8
Molekylær formel C10 H12 Cl N O
Mængde 2.5 g
Synonym Propionamide, N-benzyl-3-chloro- (6CI,7CI,8CI), Beclamid, Beclamide, Beklamid, Chloracon, Chlorakon, Chloroethylphenamide, Hibicon, N-(3-Chloropropionyl)benzylamine, N-Benzyl-3-chloropropionamide, N-Benzyl-β-chloropropanamide, N-Benzyl-β-chloropropionamide, NSC 67062, Neuracen, Nidrane, Nydran, Nydrane, Posedrine, Seclar, Propanamide, 3-chloro-N-(phenylmethyl)-
InChI formel InChI=1S/C10H12ClNO/c11-7-6-10(13)12-8-9-4-2-1-3-5-9/h1-5H,6-8H2,(H,12,13)
SMIL ClCCC(=O)NCc1ccccc1
IUPAC navn N-benzyl-3-chloropropanamide
Molekylvægt (g/mol) 197.66
Formel vægt 197.0607