missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BEC hydrochloride, MedChemExpress
Description
BEC hydrochloride is a slow-binding and competitive Arginase II inhibitor with Ki of 0.31 μM and 30 nM at pH 7.5 and pH 9.5, respectively.
Specifications
Specifications
| Kemisk navn eller materiale | BEC hydrochloride |
| Fysisk form | Powder |
| CAS | 222638-67-7 |
| Farve | Off-White |
| Mængde | 50 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C5H13BClNO4S |
| Til brug med (applikation) | Cancer-programmed cell death |
| Opløselighedsinformation | DMSO : 50 mg/mL (217.87 mM; Need ultrasonic) ∣H2O : 5 mg/mL (21.79 mM; Need ultrasonic) |
| SMIL | N[C@@H](CSCCB(O)O)C(O)=O.[H]Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?