Få mere at vide
BD Recomninant Rat GM-CSF
Beskrivelse
- Produced by activated T cells, macrophages, vascular endothelial cells and fibroblast
- Acts on mature leukocytes to promote inflammatory responses in peripheral tissues
- Promotes inflammatory responses
- Storage buffer: frozen aqueous buffered solution containing BSA and glycerol
ELISA standard, bioassay
Tekniske data
Tekniske data
| Indhold og opbevaring | -80 °C |
| Mængde | 5μ g |
| Kilde | Rotte |
Må kun bruges til undersøgelsesformål.