Få mere at vide
BD Recombinant Human Monocyte Chemoattractant Protein-3
Beskrivelse
MCP-3 Characteristics:
- Member of CC chemokine family
- Produced by monocytes, platelets, fibroblasts and osteosarcoma cell line MG-63
- Target cells include monocytes, lymphocytes, basophils, eosinophils, NK cells, dendritic cells and neutrophils
- Maximal Activity Range: 50-200 ng/ml
- ELISA standard
- Bioassay
Tekniske data
Tekniske data
| Koncentration | 200 μg/mL |
| Type | PharMingen™ Recombinant Human Monocyte Chemoattractant Protein-3 |
| Til brug med (applikation) | ELISA, FA, Standard |
| Mængde | 500 μg |
| Form | Liquid |