Få mere at vide
BD Human Th1/Th17 Phenotyping Kit
Artikelnummer. 15880459
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
15880459 1 sæt
1 options
Artikelnummer. 15880459 Leverandør BD Leverandørnr. 560752
 Begrænset vare
Forventet forsendelse: 13-08-2026
Læg i indkøbskurv
Læg i indkøbskurv
Indeholder
Human Th1/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, IFN-γ FITC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Simultaneously view Th1 and Th17 cells in mouse model systems
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
  • Can be used to successfully analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 1 sæt
For research use only. Not for diagnostic or therapeutic procedures.