Få mere at vide
BD Human T Cell Activation Kit
Artikelnummer. 15870459
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
15870459 1 sæt
1 options
Artikelnummer. 15870459 Leverandør BD Leverandørnr. 560750
 Begrænset vare
Forventet forsendelse: 28-10-2026
Indeholder
Alexa Fluor™ 647 Conjugates: p38 MAPK (pT180/Y182), ERK (pT202/204), Stat1 (pY701), Stat3 (pY705), Stat5 (pY694), and Stat6 (pY641) Human T Cell Cocktail, Compensation Reagents, Control Cells and buffers
Reliably determines the level of key phosphorylated signaling proteins involved in T-cell activation
  • Ready to use
  • Supplied with pre-titrated fluorochrome conjugated antibodies, optimized buffers, instrument setup beads, lyophilized human control cells, validated protocols
  • Includes access to Cytobank software for storing, sharing and analysis of data
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 1 sæt
For research use only. Not for diagnostic or therapeutic procedures.