Learn More
BD Difco™ Dehydrated Culture Media: Wort Agar
For cultivation and enumeration of yeasts
Brand: BD 211671
Additional Details : CAS Number : 9004-53-9 Weight : 0.80000kg
Description
- Formulation: Malt extract 15g/L, Peptone 0.78g/L, Maltose 12.75g/L, Dextrin 2.75g/L, Glycerol 2.35g/L, Dipotassium Phosphate 1g/L, Ammonium Chloride 1g/L, Agar 15g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
ISO/DIS, 11014
Specifications
ISO/DIS, 11014 | |
Dehydrated | |
Agar | |
For cultivation and enumeration of yeasts | |
500 g |
<30°C | |
Wort Agar | |
Malt extract 15g/L, Peptone 0.78g/L, Maltose 12.75g/L, Dextrin 2.75g/L, Glycerol 2.35g/L, Dipotassium Phosphate 1g/L, Ammonium Chloride 1g/L, Agar 15g/L | |
Bottle |