missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: UBA Medium
For cultivation of microorganisms significant in brewing industry
Brand: BD 285610
Additional Details : Weight : 0.80000kg
Specifications
ISO/DIS, 11014 | |
Dehydrated | |
Agar | |
Isolation of microorganisms | |
500 g |
25°C | |
Universal Beer Agar | |
Yeast extract 6.1g/L, Peptonized Milk 15g/L, Tomato Juice (from 244mL) 12.2g/L, Dextrose 16.1g/L, Dipotassium Phosphate 0.31g/L, Monopotassium Phosphate 0.31g/L, Magnesium Sulfate 0.12g/L, Sodium Chloride 6mg/L, Ferrous Sulfate 6mg/L, Manganese Sulfate 6mg/L, Agar 12g/L | |
Bottle |