missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Raka-Ray No. 3 Medium
For isolation of lactic acid bacteria encountered in beer and the brewing process
Brand: BD 218671
Additional Details : Weight : 0.80000kg
Description
- Formulation: Yeast extract 5g/L, Tryptone 20g/L, liver Concentrate 1g/L, Maltose 10g/L, Fructose 5g/L, Glucose 5g/L, Betaine Hydrochloride 2g/L, Diammonium Citrate 2g/L, Potassium Aspartate 2.5g/L, Magnesium Sulfate 0.98g/L, Manganese Sulfate 0.42g/L, Dipotassium Phosphate 2g, N-Acetyl Glucosamine 0.5g, Potassium Glutamate 2.5g, Agar 16g
Order Info
Prices are for units of one each unless otherwise specified.
Specifications
<30°C | |
Raka-Ray No. 3 | |
Yeast extract 5g/L, Tryptone 20g/L, liver Concentrate 1g/L, Maltose 10g/L, Fructose 5g/L, Glucose 5g/L, Betaine Hydrochloride 2g/L, Diammonium Citrate 2g/L, Potassium Aspartate 2.5g/L, Magnesium Sulfate 0.98g/L, Manganese Sulfate 0.42g/L, Dipotassium Phosphate 2g/L, N-Acetyl Glucosamine 0.5g/L, Potassium Glutamate 2.5g/L, Agar 16g/L | |
Bottle |
Dehydrated | |
Dehydrated Culture Media | |
For enumerating heterotrophic organisms in treated potable Water | |
500 g |