missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydreret kulturmedie: Antibiotisk medium 5

Product Code. 11792864
Change view
Click to view available options
Mængde:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
11792864 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11792864 Supplier BD Supplier No. 227710

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Til bestemmelse af antibiotisk styrke ved den mikrobiologiske analyseteknik

  • Streptomycin Assay Agar Mikrobiologisk analyse af antibiotika

TRUSTED_SUSTAINABILITY

Specifications

Certificeringer/compliance AOAC, USDA, USP
Indhold og opbevaring 2 °C til 8 °C
Form Dehydreret
Medietype Antibiotisk medium 5
Produkttype Antibiotisk medium
Formulering Oksekødekstrakt 1,5 g/l, gærekstrakt 3 g/l, pepton 6 g/l, agar 15 g/l
Til brug med (applikation) Streptomycin assay Agar Mikrobiologisk assay af antibiotika
Emballagetype Flaske
Mængde 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.