Learn More
Cytometric Bead Array Wash Buffer, BD
Product Code. 15809938 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
130mL
Unit Size:
Each
Product Code. Quantity Unit Size
15809938 130mL Each
1 options
Product Code. 15809938 Supplier BD Biosciences Supplier No. 560105
 Restricted Item
Forventet forsendelse: 03-12-2026
To be used with the BD CBA Flex Set assays
  • For wash steps and to resuspend beads for analysis
TRUSTED_SUSTAINABILITY
Specifications
Color White
Content And Storage 4°C
Format Liquid
Quantity 130mL
Validated Application 29 CFR 1910.1200, 91/155/EEC, 93/112/EC, RUO
For research use only.