Få mere at vide
BD BrdU In-Situ Detection Kit
Artikelnummer. 15876388
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
15876388 1 sæt
1 options
Highly toxic, avoid contact with skin, eyes, and mucous membranes
Artikelnummer. 15876388 Leverandør BD Leverandørnr. 550803
 Begrænset vare
Forventet forsendelse: 29-07-2026
Læg i indkøbskurv
Læg i indkøbskurv
Highly toxic, avoid contact with skin, eyes, and mucous membranes
Indeholder
Fixation Buffer, Diluent Buffer, BD™ Retrievagen A Antigen Retrieval Solution, biotinylated anti-BrdU antibody, ready-to-use Streptavidin-Horseradish peroxidase (SAv-HRP) Solution, DAB Buffer, DAB Chromogen, and Control Slides
Kræver
BrdU for in vitro or in vivo labeling, H2O2 (30%) solution for blocking endogenous peroxidase, phosphate buffer saline (PBS)
Determines proliferation rates and cell cycle kinetics in normal and cancerous tissues
Cellular incorporation of BrdU during S phase can be readily detected by anti-BrdU-specific antibodies following membrane permeabilization. Subsequent quantitation of cells positive for BrdU is possible using routine methods for cell analysis
  • Measures labeling indices of different cell populations
  • Improved specific staining with minimal background
  • Designed antigen retrieval solution unmask antigenic sites and preserve tissue morphology
  • Enables study of proliferation state of phenotypically defined cells within the micro-environment of tissues
  • Reaction: 50 slides
Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 1 sæt
Kun forskningsbrug