Learn More
BD BBL™ Dehydrated Culture Media: Thioglycollate Medium Brewer Modified
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Brand: BD 211716
Additional Details : CAS Number : 7647-14-5 Weight : 0.54000kg
Description
Cultivation of obligate anaerobes, microaerophiles and facultative organism
Certifications
CE, USP
Specifications
CE, USP | |
Dehydrated | |
Microbiology Media | |
Cultivation of obligate anaerobes, microaerophiles and facultative organism | |
500 g |
2 to 25°C | |
Thioglycollate Medium Brewer Modified | |
Pancreatic digest of Casein 17.5g/L, Papaic digest of Soybean Meal 2.5g/L, Dextrose 10g/L, Sodium Chloride 5g/L, Sodium Thioglycollate 1g/L, Dipotassium Phosphate 2g/L, Methylene blue 2mg/L, Agar 0.5g/L | |
Bottle |
Safety and Handling
- thioglycollate medium, brewer modified
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium chloride