Learn More
BD BBL™ Dehydrated Culture Media: Schaedler Agar
For several media formulations used for recovery of anaerobic microorganisms
Brand: BD 212189
This item is not returnable.
View return policy
Description
Specifications
CE | |
Dehydrated | |
Agar | |
For several media formulations used for the recovery of anaerobic microorganisms | |
500 g |
Refrigerate | |
Schaedler Agar Base | |
Pancreatic digest of Casein 8.2g/L, Peptic digest of Animal Tissue 2.5g/L, Papaic digest of Soybean Meal 1g/L, Dextrose 5.8g/L, Yeast extract 5g/L, Sodium Chloride 1.7g/L, Dipotassium Phosphate 0.8g/L, L-Cystine 0.4g/L, Hemin 0.01g/L, Tris (hydroxymethyl) aminomethane 3g/L, Agar 13.5g/L | |
Bottle |
Product Content Correction
Product Title
BD BBL™ Dehydrated Culture Media: Schaedler Agar > Schaedler Agar Base; 500g
Spot an opportunity for improvement?Share a Content Correction