Få mere at vide
BD 1063-d8 Dihydrochloride, TRC
Artikelnummer. 30338426
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30338426 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338426 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1294721MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[2-(3,4-Dichlorophenyl)ethyl]-4-methylpiperazine-d8 Dihydrochloride
CAS 1246819-69-1
Molekylær formel C13 D8 H10 Cl2 N2
Mængde 1 mg
Synonym BD 1063-d8 Dihydrochloride, 1-[2-(3,4-Dichlorophenyl)ethyl]-4-methylpiperazine-d8 Dihydrochloride
InChI formel InChI=1S/C13H18Cl2N2/c1-16-6-8-17(9-7-16)5-4-11-2-3-12(14)13(15)10-11/h2-3,10H,4-9H2,1H3/i6D2,7D2,8D2,9D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])N(CCc2ccc(Cl)c(Cl)c2)C1([2H])[2H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuterio-1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine
Molekylvægt (g/mol) 281.251
Formel vægt 280.135