Learn More
BCL2L14 Recombinant Rabbit Monoclonal Antibody (ARC2474), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Thermo Scientific MA537912
Description
Positive Samples: A-549, BxPC-3, Mouse large intestine, Mouse testis, Mouse stomach, Rat testis Immunogen sequence: MCSTSGCDLE EIPLDDDDLN TIEFKILAYY TRHHVFKSTP ALFSPKLLRT RSLSQRGLGN CSANESWTEV SWPCRNSQSS EKAINLGKKK SSWKAFFGVV EKEDSQSTPA KVSAQGQRTL EYQDSHSQQW SRCLSNVEQC LEHEAVDPKV ISIANRVAEI VYSWPPPQAT QAGGFKSKEI FVTEGLSFQL QGHVPVASSS KKDEEEQILA KIVELLKYSG DQLERKLKKD KALMGHFQDG LSYSVFKTIT
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications
BCL2L14 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYK4, Q9BZR8, Q9CPT0 | |
BCL2L14 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human BCL2L14/BCLG (Q9BZR8) | |
100 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
ARC2474 | |
Unconjugated | |
BCL2L14 | |
4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26 | |
Rabbit | |
Affinity chromatography | |
RUO | |
500348, 66813, 79370 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.