missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Antibody (BCL2L1/4509R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14165AF700
Additional Details : Weight : 0.00970kg
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
bcl-x | |
Monoclonal | |
Alexa Fluor 700 | |
Rabbit | |
Protein A or G purified | |
RUO | |
598 | |
Store at 4°C in the dark. | |
IgG κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
BCL2L1/4509R | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Recombinant fragment (around aa1-100) of human bcl-x protein (exact sequence is proprietary) (Uniprot: Q07817 ) | |
0.1 mL | |
Primary | |
Human | |
Purified |