Få mere at vide
MedChemExpress BCECF-AM
Tekniske data
Tekniske data
| Indhold og opbevaring | -20°C, protect from light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Produkttype | Dye Reagents |
| Mængde | 1 mg |
| Forsendelsestilstand | Room temperature |
| Molekylvægt (g/mol) | 820.7 g/mol |
| CAS | 117464-70-7 |