missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCCIP Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-00296AF700
Additional Details : Weight : 0.00970kg
Description
BCCIP Polyclonal specifically detects BCCIP in Human samples. It is validated for ELISA.Specifications
BCCIP | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 700 | |
BCCIPalpha, BRCA2 and CDKN1A interacting protein, BRCA2 and CDKN1A-interacting protein, BRCA2 and Cip1/p21 interacting protein, cdk inhibitor p21 binding protein, P21- and CDK-associated protein 1, Protein TOK-1, TOK-1, TOK-1alpha, TOK1BCCIPbeta, TOK-1beta | |
Produced in rabbits immunized with purified, recombinant BCCIP (AAH09771.1; Met1-Val314). | |
0.1 mL | |
Cell Cycle and Replication | |
56647 | |
Store at 4°C in the dark. | |
IgG |