missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BD BBL™ Mannitol Salt Agar
Klik for at se tilgængelige muligheder
:
Mannitol Salt Agar; Quantity: 5 lb.
Beskrivelse
- Mannitol Salt Agar is used for the selective isolation and enumeration of staphylococci from clinical and nonclinical materials.
- Mannitol Salt Agar is a nutritive medium due to its content of peptones and beef extract, which supply essential growth factors, such as nitrogen, carbon, sulfur and trace nutrients.
- Mannitol fermentation, as indicated by a change in the phenol red indicator, aids in the differentiation of staphylococcal species.
Tekniske data
Tekniske data
| Form | Dehydrated |
| Medietype | Mannitol Salt Agar |
| Formulering | Pancreatic Digest of Casein 5.0 g/L, Peptic Digest of Animal Tissue 5.0 g/L, Beef Extract 1.0 g/L, Sodium Chloride 75.0 g/L, D-Mannitol 10.0 g/L, Phenol Red 25.0 mg/L, Agar 15.0 g/L |
| Til brug med (applikation) | Selective Isolation and Enumeration of Staphylococci from Clinical and Nonclinical Materials. |
| Emballagetype | Pail |
| pH | 7.4 ± 0.2 |
| Mængde | 5 lb. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion