Få mere at vide
Bazedoxifene-d4, TRC
Artikelnummer. 30338811
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338811 1 mg
30333189 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338811 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1292471MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bazedoxifene-d4 Acetate
CAS 1133695-49-4
Molekylær formel C30 H30 D4 N2 O3
Mængde 1 mg
Synonym Bazedoxifene-d4 Acetate, 1-[[4-[2-(Hexahydro-1H-azepin-1-yl)(ethoxy-d4)]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol Acetate, TSE 424-d4, Viviant-d4, WAY-TES 424-d4
InChI formel InChI=1S/C30H34N2O3/c1-22-28-20-26(34)12-15-29(28)32(30(22)24-8-10-25(33)11-9-24)21-23-6-13-27(14-7-23)35-19-18-31-16-4-2-3-5-17-31/h6-15,20,33-34H,2-5,16-19,21H2,1H3/i18D2,19D2
SMIL [2H]C([2H])(Oc1ccc(Cn2c(c(C)c3cc(O)ccc23)c4ccc(O)cc4)cc1)C([2H])([2H])N5CCCCCC5
IUPAC navn 1-[[4-[2-(azepan-1-yl)-1,1,2,2-tetradeuterioethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methylindol-5-ol
Molekylvægt (g/mol) 474.627
Formel vægt 474.282