Få mere at vide
Bazedoxifene Acetate, LoGiCal
Artikelnummer. 30460115
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460115 10 mg
1 options
Artikelnummer. 30460115 Leverandør LoGiCal Leverandørnr. LGCFOR3342.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bazedoxifene Acetate
CAS 198481-33-3
Analyt- eller komponentnavne Bazedoxifene Acetate
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C30 H34 N2 O3 . C2 H4 O2
Certificeringer/compliance ISO 9001
Synonym 1H-Indol-5-ol, 1-[[4-[2-(hexahydro-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-, acetate (1:1), 1H-Indol-5-ol, 1-[[4-[2-(hexahydro-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-, monoacetate (salt) (9CI), Bazedoxifene acetate, TSE 424, Viviant, WAY-TES 424
InChI formel InChI=1S/C30H34N2O3.C2H4O2/c1-22-28-20-26(34)12-15-29(28)32(30(22)24-8-10-25(33)11-9-24)21-23-6-13-27(14-7-23)35-19-18-31-16-4-2-3-5-17-31;1-2(3)4/h6-15,20,33-34H,2-5,16-19,21H2,1H3;1H3,(H,3,4)
Løsningstype Neat
SMIL CC1=C(N(C2=C1C=C(O)C=C2)CC3=CC=C(OCCN4CCCCCC4)C=C3)C5=CC=C(O)C=C5.CC(O)=O
IUPAC navn acetic acid;1-[[4-[2-(azepan-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methylindol-5-ol
Industry Type Clinical Testing
Molekylvægt (g/mol) 530.65
Mængde 10 mg
Formel vægt 530.2781 g/mol