Få mere at vide
BAY-876, TRC
Artikelnummer. 30180180
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180180 5 mg
30179821 1 mg
30180064 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180180 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1275305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1799753-84-6
Molekylær formel C24H16F4N6O2
Molekylvægt (g/mol) 496.42
Synonym BAY-876, N4-[1-[(4-Cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-quinolinedicarboxamide
IUPAC navn 4-N-[1-[(4-cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)pyrazol-4-yl]-7-fluoroquinoline-2,4-dicarboxamide
SMIL Cc1c(NC(=O)c2cc(nc3cc(F)ccc23)C(=O)N)c(nn1Cc4ccc(cc4)C#N)C(F)(F)F
Tekniske data
Kemisk navn eller materiale N4-[1-[(4-Cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-quinolinedicarboxamide
Mængde 5 mg
InChI formel InChI=1S/C24H16F4N6O2/c1-12-20(21(24(26,27)28)33-34(12)11-14-4-2-13(10-29)3-5-14)32-23(36)17-9-19(22(30)35)31-18-8-15(25)6-7-16(17)18/h2-9H,11H2,1H3,(H2,30,35)(H,32,36)
Formel vægt 496.1271