Få mere at vide
BAY 678, Tocris Bioscience™
Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor
Brand: Tocris Bioscience 5706/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BAY 678 is a potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor (IC50 = 20 nM). Exhibits around 2000-fold selectivity for NHE against a panel of 21 other serine proteases. Effective in an acute lung injury model in vivo. Orally bioavailable. Inactive S-enantiomer (Cat.No.6389) also available.Tekniske data
| 5-[(4R)-5-Acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-2-pyridinecarbonitrile | |
| 675103-36-3 | |
| >98% | |
| 5 mg |