missing translation for 'onlineSavingsMsg'
Få mere at vide

BAY-678, MedChemExpress

Artikelnummer. 16494458
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16494458 10 mg
16463096 25 mg
16437187 5 mg
16430777 50 mg
16415346 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16494458 Leverandør Medchem Express Leverandørnr. HY111457A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).

BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).

Specifications

Kemisk navn eller materiale BAY-678
Fysisk form Solid
CAS 675103-36-3
Farve Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C20H15F3N4O2
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : ≥ 100 mg/mL (249.78 mM) ∣Ethanol : ≥ 4.76 mg/mL (11.89 mM)
SMIL N#CC1=NC=C([C@H](C(C(C)=O)=C(C)N2C3=CC=CC(C(F)(F)F)=C3)NC2=O)C=C1
Molekylvægt (g/mol) 400.35
Formel vægt 400.35
Procent renhed 99.24%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.