Få mere at vide
Tocris Bioscience™ BAY 6096
Beskrivelse
BAY 6096 is a potent (IC50 = 14 nM), and selective adrenergic α2B receptor antagonist that displays 394, 725, >845-fold selectivity over α1A, α2A, and α2C, respectively. BAY 6096 reduces α2B receptor agonist-induced vasoconstriction and has a high clearance and low mean residence times in rats. BAY 6096 is highly water-soluble.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-[2-[[[3-(3,5-Dimethyl-4-isoxazolyl)imidazo[1,2-a]pyridin-7-yl]carbonyl]amino]ethyl]-4-(methylamino)pyridinium chloride |
| CAS | 2313483-41-7 |
| Mængde | 50 mg |
| Mål | Adrenergic alpha-2 Receptor Antagonists |
| Molekylær formel | C21H23ClN6O2 |
| Renhed | 0.98 |