missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bay 41-4109, MedChemExpress
Description
BAY 41-4109 is a potent inhibitor of human hepatitis B virus (HBV) with an IC50 of 53 nM.
Specifications
Specifications
| Kemisk navn eller materiale | Bay 41-4109 |
| CAS | 298708-81-3 |
| Farve | Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C18H13ClF3N3O2 |
| Mængde | 100 mg |
| Opløselighedsinformation | DMSO : 100 mg/mL (252.68 mM; Need ultrasonic) |
| SMIL | O=C(C1=C(C)N=C(C2=NC=C(F)C=C2F)N[C@H]1C3=CC=C(F)C=C3Cl)OC |
| Molekylvægt (g/mol) | 395.76 |
| Formel vægt | 395.76 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?