Få mere at vide
Tocris™ BAY 293
Beskrivelse
BAY 293 is a potent KRas/son of sevenless 1 (SOS1) interaction inhibitor (IC50 = 21 nM); active R-enantiomer. Downregulates active RAS in tumor cells. Inhibits RAS-RAF-MEK-ERK pathway. Exhibits synergistic effects with KRASG12C inhibitor ARS-853.
Negative control also available.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (R)-6,7-Dimethoxy-2-methyl-N-[1-[4-[2-[(methylamino)methyl]phenyl]thiophene-2-yl]ethyl]quinazolin-4-amine |
| CAS | 2244904-70-7 |
| Mængde | 5 mg |
| Molekylær formel | C25H28N4O2S |
| Renhed | 98% |