Learn More
Fisherbrand™ Basket Inserts for Elmasonic X-tra ST Ultrasonic Cleaners
Place bulk material on these basket inserts for thorough ultrasonic cleaning.
Brand: Fisherbrand™
Additional Details : Weight : 6.00000kg
Description
- Mesh construction
- Available in a range of sizes for all X-tra ST models
- Accessory item
Specifications
Basket Insert | |
Ultrasonic cleaning | |
7 x 7 x 1 mm |
Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H | |
Wire Mesh |