Learn More
Barium Hydroxide Octahydrate, ACS Reagent, ≥98%, Honeywell™ Fluka™
Catalog Number 15642060
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
12230-71-6
BaH18O10
315.46
MFCD00149152
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
17749109
barium(2+);dihydroxide;octahydrate
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
12230-71-6
BaH18O10
315.46
MFCD00149152
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
17749109
barium(2+);dihydroxide;octahydrate
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
Melting Point 78°C
Color Colorless
Physical Form Solid
Quantity 2.5 kg
Linear Formula Ba(OH)2 · 8H2O
UN Number UN3262
Formula Weight 315.46g/mol
Percent Purity ≥98%
Grade ACS Reagent
Packaging Poly Bottle
Density 2180g/cu.cm
Chemical Name or Material Barium Hydroxide Octahydrate