Learn More
Cole-Parmer™ BarbLock Assemblies
Provide uniform 360-degree radial crimping for a leak-free, durable seal. Cole-Parmer™ Barblock Assemblies feature an easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage.
Brand: Cole-Parmer™ 30604-10
Additional Details : Weight : 0.45000kg
Description
- Safe to use with pressures up to 140psi or upper pressure limit of tubing
- An excellent alternative to securing tubing and hose barbs with cable ties — no more crimps or tube distortion
- BarbLock assembly consists of three pieces: the sleeve, collect, and barbed fittings
- To install the fitting, slide the sleeve and collet onto the tubing and then inserts the barbed fitting into the tubing
- Prelock the sleeve and collet by pressing them together until you hear a click
- Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool
- Once assembled, the BarbLock assembly can only be unlocked with the removal tool
Specifications
BarbLock Assemblies | |
Fits 1/4 in. and L/S 15, 17, & 24 Tubing | |
Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage | |
1 Pack |