Få mere at vide
Bamipine Hydrochloride, LoGiCal
Artikelnummer. 30459678
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30459678 5 mg
1 options
Artikelnummer. 30459678 Leverandør LoGiCal Leverandørnr. LGCFOR1824.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bamipine Hydrochloride
CAS 1229-69-2
Analyt- eller komponentnavne Bamipine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H24 N2 . Cl H
Certificeringer/compliance ISO 9001
Synonym 4-Piperidinamine, 1-methyl-N-phenyl-N-(phenylmethyl)-, monohydrochloride (9CI), Piperidine, 4-(N-benzylanilino)-1-methyl-, hydrochloride (7CI,8CI), 4-Piperidinamine, 1-methyl-N-phenyl-N-(phenylmethyl)-, hydrochloride (1:1), Bamipine Hydrochloride
InChI formel InChI=1S/C19H24N2.ClH/c1-20-14-12-19(13-15-20)21(18-10-6-3-7-11-18)16-17-8-4-2-5-9-17;/h2-11,19H,12-16H2,1H3;1H
Løsningstype Neat
SMIL Cl.CN1CCC(CC1)N(Cc2ccccc2)c3ccccc3
IUPAC navn N-benzyl-1-methyl-N-phenylpiperidin-4-amine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 316.87
Mængde 5 mg
Formel vægt 316.1706 g/mol