missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bamifylline Hydrochloride, Mikromol™
Beskrivelse
Bamifylline Hydrochloride
Specifications
Specifications
| Kemisk navn eller materiale | Bamifylline Hydrochloride |
| CAS | 20684-06-4 |
| Analyt- eller komponentnavne | Bamifylline Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H27 N5 O3 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 1H-Purine-2,6-dione, 7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-3,7-dihydro-1,3-dimethyl-8-(phenylmethyl)-, monohydrochloride (9CI), Theophylline, 8-benzyl-7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-, monohydrochloride (8CI), AC 3810, BAX 2739Z, Bamifylline hydrochloride, Bamifylline monohydrochloride, Benzethamophylline hydrochloride, Briofil, Trentadil, 1H-Purine-2,6-dione, 7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-3,7-dihydro-1,3-dimethyl-8-(phenylmethyl)-, hydrochloride (1:1), Bamifylline Hydrochloride |
| InChI formel | InChI=1S/C20H27N5O3.ClH/c1-4-24(12-13-26)10-11-25-16(14-15-8-6-5-7-9-15)21-18-17(25)19(27)23(3)20(28)22(18)2;/h5-9,26H,4,10-14H2,1-3H3;1H |
| SMIL | Cl.CCN(CCO)CCn1c(Cc2ccccc2)nc3N(C)C(=O)N(C)C(=O)c13 |
| IUPAC navn | 8-benzyl-7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-1,3-dimethylpurine-2,6-dione;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?