Få mere at vide
Bamifylline Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bamifylline Hydrochloride |
| CAS | 20684-06-4 |
| Analyt- eller komponentnavne | Bamifylline Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H27 N5 O3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1H-Purine-2,6-dione, 7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-3,7-dihydro-1,3-dimethyl-8-(phenylmethyl)-, monohydrochloride (9CI), Theophylline, 8-benzyl-7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-, monohydrochloride (8CI), AC 3810, BAX 2739Z, Bamifylline hydrochloride, Bamifylline monohydrochloride, Benzethamophylline hydrochloride, Briofil, Trentadil, 1H-Purine-2,6-dione, 7-[2-[ethyl(2-hydroxyethyl)amino]ethyl]-3,7-dihydro-1,3-dimethyl-8-(phenylmethyl)-, hydrochloride (1:1), Bamifylline Hydrochloride |
| InChI formel | InChI=1S/C20H27N5O3.ClH/c1-4-24(12-13-26)10-11-25-16(14-15-8-6-5-7-9-15)21-18-17(25)19(27)23(3)20(28)22(18)2;/h5-9,26H,4,10-14H2,1-3H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCN(CCO)CCn1c(Cc2ccccc2)nc3N(C)C(=O)N(C)C(=O)c13 |
| Vis mere |