Få mere at vide
Tocris Bioscience™ BAM 15
Mitochondrial protonophore uncoupler
Brand: Tocris Bioscience™ 5737/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BAM 15 is a mitochondrial protonophore uncoupler; uncouples oxidative phosphorylation in L6 myoblast mitochondria in vitro (EC50 = 270 nM). Does not depolarize the plasma membrane. Protects mice from acute renal ischemic-reperfusion injury.Tekniske data
| N5,N6-bis(2-Fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine | |
| 210302-17-3 | |
| 50 mg |