missing translation for 'onlineSavingsMsg'
Få mere at vide

Baloxavir, MedChemExpress

Artikelnummer. 16485186
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16485186 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16485186 Leverandør Medchem Express Leverandørnr. HY109025A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Baloxavir (Baloxavir acid), derived from the prodrug Baloxavir marboxil, is a first-in-class, potent and selective cap-dependent endonuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses. Baloxavir inhibits viral RNA transcription and replication and has potently antiviral activity.

Baloxavir (Baloxavir acid), derived from the prodrug Baloxavir marboxil, is a first-in-class, potent and selective cap-dependent endonuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses. Baloxavir inhibits viral RNA transcription and replication and has potently antiviral activity.

Tekniske data

Kemisk navn eller materiale Baloxavir
Fysisk form Powder
CAS 1985605-59-1
Farve Off-White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C24H19F2N3O4S
Til brug med (applikation) COVID-19-anti-virus
Synonym Baloxavir acid S-033447
Opløselighedsinformation DMSO : 41.67 mg/mL (86.19 mM; Need ultrasonic)
SMIL O=C1N2[C@](COCC2)([H])N([C@@H]3C4=CC=CC=C4SCC5=C(F)C(F)=CC=C35)N6C1=C(O)C(C=C6)=O
Molekylvægt (g/mol) 483.49
Formel vægt 483.49
Procent renhed 98.92%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.