Learn More
Lenz Laborglasinstrumente™ Ball Members, Inch Range
Manufactured from DURAN™ tubing. Lenz Laborglasinstrumente™ Ball Members, Inch Range are perfect for mixing solutions in a centrifuge tube.
192.92 DKK - 288.32 DKK
Product Code | Brand | Overall Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Overall Length (Metric) | Price | Quantity & Availability | |||||
10040201
|
Lenz Laborglasinstrumente™
01215130 |
150 mm |
288.32 DKK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10253951
|
Lenz Laborglasinstrumente™
01214125 |
120 mm |
192.92 DKK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN 12264 and in addition to standard
- Made of DURAN™ tubing
Specifications
Borosilicate Glass | |
1 |