missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAK Recombinant Rabbit Monoclonal Antibody (7Q3T8), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Thermo Scientific MA537832
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, AGS Immunogen sequence: RQECGEPALP SASEEQVAQD TEEVFRSYVF YRHQQEQEAE GVAAPADPEM VTLPLQPSST MGQVGRQLAI IG
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress.
Specifications
BAK | |
Recombinant Monoclonal | |
0.75 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q16611 | |
BAK1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 11-82 of human Bak (NP_0011791) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
7Q3T8 | |
Unconjugated | |
BAK1 | |
Apoptosis regulator BAK; BAK; Bak1; BAK-LIKE; BCL2 antagonist/killer 1; Bcl2 homologous antagonist/killer; Bcl-2 homologous antagonist/killer; BCL2-antagonist/killer 1; BCL2L7; bcl2-L-7; BCL2-like 7 protein; BCL2-like 7protein; bcl-2-like protein 7; Bcl-2-likeous antagonist/killer; CDN1; MGC117255; MGC3887; N-Bak; N-BAK1; pro-apoptotic protein BAK | |
Rabbit | |
Affinity chromatography | |
RUO | |
578 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BAK Recombinant Rabbit Monoclonal Antibody (7Q3T8), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction