Få mere at vide
R&D Systems™ BAK BH3 Synthetic Peptide GQVGRQLAIIGDDINR Protein
Mængde:
100 μg
Beskrivelse
The BAK BH3 Synthetic Peptide GQVGRQLAIIGDDINR Protein has been validated for the following applications: Inhibition Activity.
Tekniske data
Tekniske data
| Gen-id (Entrez) | 578 |
| Molekylvægt (g/mol) | 1.7 kDa |
| Navn | BAK BH3 Domain, aa 72-87 |
| Mængde | 100 μg |
| Opbevaringskrav | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Renhed eller kvalitetsklasse | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |