Få mere at vide
Baicalein Monohydrate, TRC
Artikelnummer. 30331169
Skift visning
Mængde:
500 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30331169 500 mg
30339219 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30331169 Leverandør Toronto Research Chemicals Leverandørnr. TRCB115250500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Baicalein, Monohydrate
CAS 352000-07-8
Molekylær formel C15H12O6
Mængde 500 mg
Synonym 5,6,7-Trihydroxyflavone Monohydrate, 5,6,7-Trihydroxy-2-phenyl-4H-1-benzopyran-4-one Hydrate
InChI formel InChI=1S/C15H10O5.H2O/c16-9-6-11(8-4-2-1-3-5-8)20-12-7-10(17)14(18)15(19)13(9)12;/h1-7,17-19H;1H2
SMIL O.Oc1cc2OC(=CC(=O)c2c(O)c1O)c3ccccc3
IUPAC navn 5,6,7-trihydroxy-2-phenylchromen-4-one;hydrate
Molekylvægt (g/mol) 288.26
Formel vægt 288.0634