missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ BAIAP2 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Brand:  Novus Biologicals™ NBP1-88711PEP

Product Code. 18310511

  • 1930.00 DKK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human BAIAP2. The BAIAP2 Recombinant Protein Antigen is derived from E. coli. The BAIAP2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP1-88711. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10458
Chromatography
0.5mg/mL
PBS and 1M Urea, pH 7.4.
BAIAP2
27kDa
0.1 mL
E.Coli
WFPFSYTRVLDSDGSDRLHMSLQQGKSSSTGNLLDKDDLAIPPPDYGAASRAFPAQTASGFKQRPYSVAVPAFSQGLDDYGARSMSRNP
Human
>80%
Store at -20°C. Avoid freeze-thaw cycles.
Blocking/Neutralizing, Control
Unlabeled
BAIAP2
RUO
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-88711. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ BAIAP2 Recombinant Protein Antigen > 0.1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only