missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BAG3 Rabbit anti-Human, Mouse, Rat, Clone: S01-3F1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-14910-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BAG3 Monoclonal antibody specifically detects BAG3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
| BAG3 | |
| Monoclonal | |
| Unconjugated | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S01-3F1 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
| BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307 | |
| Recombinant protein of human BAG3 (Uniprot#: O95817) | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Endocrinology | |
| 9531 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion