missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (BAFFR/1557) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08867
Additional Details : Weight : 0.00970kg
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
BAFFR/TNFRSF13C | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
BAFFR/1557 | |
Unconjugated | |
10mM PBS | |
Recombinant full-length human BAFFR/TNFRSF13C protein | |
100 μg | |
Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
115650 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |