missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Bacto™ CD Supreme Fermentation Production Medium Plus, 1 L

Artikelnummer. 31781078
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31781078 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31781078 Leverandør Gibco™ Leverandørnr. A9378401

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Chemically defined medium optimized for E. coli plasmid DNA and recombinant protein production.

Gibco Bacto CD Supreme Fermentation Production Medium (FPM) Plus is the next-generation, chemically defined formulation designed to support a wide range of Escherichia coli (E. coli) strains used in plasmid DNA and recombinant protein production. This hydrolysate-free medium enhances microbial culture performance in both batch and fed-batch fermentation processes, offering superior consistency and flexibility across manufacturing scales.

Features of Bacto CD Supreme FPM Plus include:

  • Consistency with a 100% chemically defined, hydrolysate-free formulation that minimizes lot-to-lot variability
  • Expanded strain compatibility to include E. coli strains such as DH10B, BL21(DE3), NEB™ Stable, Stbl2, Stbl3, JM109, and other auxotrophic variants
  • Simplified workflows using a one-part, ready-to-use formulation that eliminates manual media preparation
  • Easy adaptation to your process with format options and compatibility with filtration or autoclave sterilization methods
  • Reduced regulatory risk with a formulation free from raw materials of animal origin or materials produced using animal-derived components

Bacto CD Supreme FPM Plus is engineered to deliver reliable and reproducible growth performance across diverse E. coli host strains, including Thi-1 mutant and auxotrophic variants. Its chemically defined composition helps ensure optimal nutrient availability without the variability introduced by hydrolysates. The medium supports high-density cultures in both batch and fed-batch modes, enabling efficient plasmid DNA or recombinant protein yield.

For fed-batch processes Bacto CD Supreme FPM Plus can be combined with Bacto CD Supreme Feed (Cat. Nos. A9378201, A9378301, A9378302) to support increased biomass accumulation and yields

This one-part, easy-to-use formulation integrates smoothly into existing manufacturing workflows and is compatible with standard sterilization methods. It is produced without any materials of animal origin, minimizing regulatory complexity and eliminating TSE/BSE risk concerns.

TRUSTED_SUSTAINABILITY

Tekniske data

Koncentration 1x
Indhold og opbevaring Store in the dark at +2°C to +8°C
Form Liquid
Fremstillingsoprindelse United States
Produkttype Culture Medium
Sterilitet Sterile
Målorganismeklasse E. coli
Test Type Microbial Cultivation
Formulering Animal Origin-free, Chemically-defined
Til brug med (applikation) Microbial Fermentation
Produktlinje Bacto CD Supreme
Mængde 1 L
Vis mere Vis mindre

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.