missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Bacto™ CD Supreme Feed (2X), liquid

Artikelnummer. 31781021
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31781021 500 mL
31781189 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31781021 Leverandør Gibco™ Leverandørnr. A9378201

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Chemically defined nutrient feed for optimized E. coli plasmid DNA and protein production.

Gibco Bacto CD Supreme Feed (2X) is an animal origin–free, chemically defined microbial feed specifically formulated to extend culture performance and increase productivity in Escherichia coli (E. coli) fed-batch processes. Designed to deliver a balanced nutrient supply, this hydrolysate-free formulation supports robust cell growth, higher biomass accumulation, and improved product yield across a broad range of E. coli expression strains.

Features of Bacto CD Supreme Feed include:

  • Increased productivity with 10-fold higher plasmid DNA production compared to batch mode
  • Consistency with a 100% chemically defined, hydrolysate-free composition that minimizes variability
  • Streamlined operations using a one-part, ready-to-use formulation that eliminates the need for complex feed preparation
  • Easy integration with Bacto CD Supreme FPM and standard sterilization methods for scalable implementation
  • Simplified compliance with an animal origin–free formulation that supports global regulatory requirements

Developed for compatibility with both Bacto CD Supreme Fermentation Production Medium (FPM) Plus (Cat. Nos. A9378401, A9378501, A9378502) and Bacto CD Supreme FPM (Cat. Nos. A4973701, A4973702), Bacto CD Supreme Feed provides sustained nutrient delivery during fed-batch fermentation that increases biomass accumulation, extends bioreactor runs, and enhances plasmid DNA and protein yield.

As a chemically defined formulation, Bacto CD Supreme Feed eliminates the performance variability associated with peptone-based feeds and supports consistent outcomes across diverse E. coli strains used in plasmid DNA and recombinant protein production.

Offered in convenient liquid and dry powder (DPM) formats, Bacto CD Supreme Feed ensures ease of use, scalability, and reliability from development to large-scale manufacturing.

TRUSTED_SUSTAINABILITY

Tekniske data

Koncentration 2x
Indhold og opbevaring Store in the dark at +2°C to +8°C
Form Liquid
Fremstillingsoprindelse United States
Produkttype E. coli Feed
Sterilitet Sterile
Målorganismeklasse E. coli
Test Type Microbial Cultivation
Formulering Animal Origin-free, Chemically-defined
Til brug med (applikation) Microbial Fermentation
Produktlinje Bacto CD Supreme
Mængde 500 mL
Vis mere Vis mindre

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.