Få mere at vide
Gibco™ Bacto™ Hjernehjerteinfusion, svin
Artikelnummer. 16239871
Skift visning
Mængde:
10 kg
Artikelnummer. Mængde
16239871 10 kg
1 options
Artikelnummer. 16239871 Leverandør Gibco™ Leverandørnr. 256110
Bacto Brain Heart Infusion, porcine, is used for cultivating a wide variety of microorganisms.
Bacto Brain Heart Infusion, porcine, is used for cultivating a wide variety of microorganisms. It was developed as an alternative to the classical Brain Heart Infusion formula and replaces calf brain and beef heart with pork brain and heart. Bacto Brain Heart Infusion, porcine, was formulated with no bovine components to minimize bovine spongiform encephalopathy (BSE) risk.
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring Opbevaringsforhold: 2 °C til 25 °C. Opbevares køligt og tørt i velforseglede beholdere.
Produktlinje Bacto
Mængde 10 kg