missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bacteriological Agar, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Description
Used extensively as a gelling agent in the preparation of bacteriological culture media. Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Superior transparency
- Reliable reproducibility
- Absence of inhibitors capable of hindering optimal development of microorganisms
- Extraneous matter reduced to minimum
- Format: White to light cream-colored powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Specifications
Color | White to light cream-colored |
Form | Powder |
Grade | Ultrapure |
Product Type | Agar |
For Use With (Application) | Used as a Gelling Agent in Preparation of Bacteriological Culture Media. |
Packaging Type | Plastic Bottle |
Quantity | 1 lb. |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'