Learn More
Bacteriological Agar, Ultrapure, Thermo Scientific Chemicals
Used as a gelling agent in preparation of bacteriological culture media.
Brand: Thermo Scientific J10906.Q1
Additional Details : CAS Number : 9002-18-0 Weight : 1.00000kg
Description
Used extensively as a gelling agent in the preparation of bacteriological culture media. Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Superior transparency
- Reliable reproducibility
- Absence of inhibitors capable of hindering optimal development of microorganisms
- Extraneous matter reduced to minimum
- Format: White to light cream-colored powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Specifications
White to light cream-colored | |
Ultrapure | |
Used as a Gelling Agent in Preparation of Bacteriological Culture Media. | |
25 lb. |
Powder | |
Agar | |
Plastic Bottle |
RUO – Research Use Only